Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11131

Sigma-Aldrich

Sodium thioantimonate(V) nonahydrate

22.5% Sb basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na3SbS4 · 9H2O
CAS Number:
Molecular Weight:
481.13
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

concentration

22.5% Sb

anion traces

chloride (Cl-): 100 mg/kg

cation traces

Fe: 50 mg/kg

SMILES string

O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[S-][Sb]([S-])([S-])=S

pictograms

Exclamation markEnvironment

signalword

Warning

pcodes

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service