Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

10400

Sigma-Aldrich

Aniline

puriss. p.a., ACS reagent, ≥99.5% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NH2
CAS Number:
Molecular Weight:
93.13
Beilstein/REAXYS Number:
605631
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

ACS reagent
puriss. p.a.

vapor density

3.22 (185 °C, vs air)

vapor pressure

0.7 mmHg ( 25 °C)

assay

≥99.5% (GC)

autoignition temp.

1139 °F

expl. lim.

11 %

impurities

hydrocarbons, in accordance
≤0.003% nitrobenzene (C6H5NO2)
≤0.1% water

ign. residue

≤0.005%

refractive index

n20/D 1.586 (lit.)
n20/D 1.586

bp

184 °C (lit.)

mp

−6 °C (lit.)

density

1.022 g/mL at 25 °C (lit.)

cation traces

Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Zn: ≤1 mg/kg

SMILES string

Nc1ccccc1

InChI

1S/C6H7N/c7-6-4-2-1-3-5-6/h1-5H,7H2

InChI key

PAYRUJLWNCNPSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

The acute toxicity of aniline involves its activation in vivo to 4-hydroxyaniline and the formation of adducts with hemoglobin. In erythrocytes, this is associated with the release of iron and the accumulation of methemoglobin and the development of hemolytic anemia and inflammation of the spleen. Tumor formation is often observed in the spleen on prolonged administration.

Physical properties

Material darkens in storage to reddish-brown with no loss in purity.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1 - STOT RE 1

target_organs

Blood

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

158.0 °F - closed cup

flash_point_c

70 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriella Fabian et al.
International journal of molecular sciences, 12(9), 6116-6134 (2011-10-22)
Toxicogenomics, based on the temporal effects of drugs on gene expression, is able to predict toxic effects earlier than traditional technologies by analyzing changes in genomic biomarkers that could precede subsequent protein translation and initiation of histological organ damage. In
Joanna Romanowska et al.
Journal of medicinal chemistry, 54(19), 6482-6491 (2011-08-13)
New synthetic protocol for the preparation of nucleoside 5'-(N-aryl)phosphoramidate monoesters 4 was developed. It consisted of a condensation of the corresponding nucleoside 5'-H-phosphonates with aromatic- or heteroaromatic amines promoted by diphenyl phosphorochloridate, followed by oxidation of the produced H-phosphonamidates with
Sylvie Mavel et al.
Bioorganic & medicinal chemistry, 20(4), 1388-1395 (2012-02-04)
The dopamine transporter (DAT) plays a pivotal role in the regulation of dopamine neurotransmission, and is involved in a number of physiological functions and brain disorders. Furthermore the DAT analysis by molecular imaging techniques is a useful tool for the
The many roles for fluorine in medicinal chemistry.
William K Hagmann
Journal of medicinal chemistry, 51(15), 4359-4369 (2008-06-24)
Lourdes Santana et al.
Journal of medicinal chemistry, 51(21), 6740-6751 (2008-10-07)
The work provides a new model for the prediction of the MAO-A and -B inhibitor activity by the use of combined complex networks and QSAR methodologies. On the basis of the obtained model, we prepared and assayed 33 coumarin derivatives

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service