Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04270

Sigma-Aldrich

Sodium phosphate monobasic

puriss., meets analytical specification of BP, anhydrous, 98-100.5% (calc. to the dried substance)

Synonym(s):

Monosodium dihydrogen orthophosphate, Monosodium phosphate, Sodium dihydrogen phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaH2PO4
CAS Number:
Molecular Weight:
119.98
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38070208
PubChem Substance ID:

grade

puriss.

assay

98-100.5% (calc. to the dried substance)

quality

meets analytical specification of BP

impurities

≤0.001% heavy metals (as Pb)

loss

≤1% loss on drying, 130 °C

pH

4.2-4.5 (20 °C, 5%)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤300 mg/kg

cation traces

As: ≤2 mg/kg
Fe: ≤10 mg/kg

SMILES string

[Na+].OP(O)([O-])=O

InChI

1S/Na.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

AJPJDKMHJJGVTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Austin C Lowder et al.
Meat science, 94(1), 9-18 (2013-01-29)
This study was conducted using response surface methodology to predict fluid loss from injected beef strip steaks as influenced by levels of salt and sodium phosphates (SP) in the injection brine. Also, a beef-based dehydrated beef protein (DBP) water binding
Meital Aviv-Gavriel et al.
Langmuir : the ACS journal of surfaces and colloids, 29(2), 683-689 (2012-12-13)
Cartilage and/or bone tissue engineering is a very challenging area in modern medicine. Since cartilage is an avascular tissue with limited capacity for self-repair, using scaffolds provides a promising option for the repair of severe cartilage damage caused by trauma
Jonathan Belsey et al.
Current medical research and opinion, 28(12), 1883-1890 (2012-11-10)
It is unclear whether small bowel visibility in video capsule endoscopy (VCE) is enhanced by the use of bowel preparation in addition to pre-procedural fasting. The objective of this study was to clarify this issue by means of a systematic
Kaisorn L Chaichana et al.
Journal of neuroscience methods, 180(1), 116-125 (2009-05-12)
The discoveries of neural (NSCs) and brain tumor stem cells (BTSCs) in the adult human brain and in brain tumors, respectively, have led to a new era in neuroscience research. These cells represent novel approaches to studying normal phenomena such
Samuel Adler et al.
Gastrointestinal endoscopy, 76(6), 1170-1174 (2012-10-03)
Colon capsule endoscopy (CCE) is a noninvasive technique for the detection of colorectal lesions. However, for CCE to be offered as an out-of-clinic procedure, the system needs to automatically alert the patient when to ingest the laxative (booster). We tested

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service