04114
Phosphorous acid
≥98.5%
Synonym(s):
Phosphonic acid
About This Item
assay
≥98.5%
impurities
≤0.002% heavy metals (as Pb)
≤1% phosphoric acid (H3PO4)
density
1.651 g/mL at 25 °C (lit.)
anion traces
chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤10 mg/kg
cation traces
Fe: ≤5 mg/kg
SMILES string
[H]P(O)(O)=O
InChI
1S/H3O3P/c1-4(2)3/h4H,(H2,1,2,3)
InChI key
ABLZXFCXXLZCGV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service