Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01068

Sigma-Aldrich

Hydrofluoric acid

technical, 51-55%

Synonym(s):

HF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HF
CAS Number:
Molecular Weight:
20.01
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

vapor density

1.27 (vs air)

vapor pressure

25 mmHg ( 20 °C)

grade

technical

assay

51-55%

impurities

≤0.02% hexafluorosilic acid (H2SiF6)

density

1.15 g/mL at 25 °C (lit.)

anion traces

sulfate (SO42-): ≤100 mg/kg

SMILES string

[H+].[F-]

InChI

1S/FH/h1H

InChI key

KRHYYFGTRYWZRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1A

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qing Wang et al.
Organic & biomolecular chemistry, 10(43), 8566-8569 (2012-09-26)
A convenient, metal-free intramolecular aminofluorination of alkenes has been developed. Employing readily available PhI(OPiv)(2) and hydrogen fluoride-pyridine in the presence of BF(3)·OEt(2), tosyl-protected pent-4-en-1-amines were converted to 3-F-piperidines in one step in good yields as well as high stereoselectivity.
Health care plan for hydrogen fluoride spill, Gumi, Korea.
Hyun-Sul Lim et al.
Journal of Korean medical science, 27(11), 1283-1284 (2012-11-21)
Angelika Baranowska-Łączkowska et al.
Journal of computational chemistry, 34(4), 275-283 (2012-09-22)
Interaction-induced static electric properties, that is, dipole moment, polarizability, and first hyperpolarizability, of the CO-(HF)(n) and N(2)-(HF)(n), n = 1-9 hydrogen-bonded complexes are evaluated within the finite field approach using the Hartree-Fock, density functional theory, Møller-Plesset second-order perturbation theory, and
Neutralizing a caustic chemical.
Santiago Nogué et al.
Nursing, 43(6), 8-8 (2013-05-21)
Peter Dinér
The journal of physical chemistry. A, 116(40), 9979-9984 (2012-09-25)
The carbonium ion has been suggested to be the intermediate in superacid-promoted reactions (SbF(5)-HF) such as hydrogen-deuterium exchange and in the electrophilic C-H cleavage into hydrogen and the carbenium ion. In this study, the superacid-promoted C-H cleavage into hydrogen and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service