Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37-0550

Sigma-Aldrich

Zinc sulfate heptahydrate

JIS special grade, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnSO4 · 7H2O
CAS Number:
Molecular Weight:
287.56
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
≥99.5%
grade:
JIS special grade
form:
solid

grade

JIS special grade

assay

≥99.5%

form

solid

reaction suitability

reagent type: catalyst
core: zinc

availability

available only in Japan

pH

4.0-6.0 (20 °C, 50 g/L)

storage temp.

2-8°C

SMILES string

O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.7H2O.Zn/c1-5(2,3)4;;;;;;;;/h(H2,1,2,3,4);7*1H2;/q;;;;;;;;+2/p-2

InChI key

RZLVQBNCHSJZPX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jorge L Rosado et al.
Food and nutrition bulletin, 33(4), 261-266 (2013-02-22)
Corn tortilla is the staple food of Mexico and its fortification with zinc, iron, and other micronutrients is intended to reduce micronutrient deficiencies. However, no studies have been performed to determine the relative amount of zinc absorbed from the fortified
Gaia Dell'Ariccia et al.
The Journal of experimental biology, 216(Pt 8), 1430-1433 (2013-01-12)
Olfactory cues have been shown to be important to homing petrels at night, but apparently those procellariiform species that also come back to the colony during the day are not impaired by smell deprivation. However, the nycthemeral distribution of homing
Mahmoud Hussein Hadwan et al.
BMC urology, 12, 32-32 (2012-11-14)
Zinc in human seminal plasma is divided into three types of ligands which are high (HMW), intermediate (IMW), and low molecular weight ligands (LMW). The present study was aimed to study the effect of Zn supplementation on the quantitative and
Laurent Dortet et al.
Antimicrobial agents and chemotherapy, 56(12), 6437-6440 (2012-10-17)
A biochemical test (Carba NP test II) was developed to identify carbapenemase production in Enterobacteriaceae and Pseudomonas spp. and to discriminate between the different types of carbapenemases (classes A, B, and D). It is based on the detection of the
O I Smian et al.
Likars'ka sprava, (3-4)(3-4), 67-71 (2013-01-30)
The article analyzed 60 patients with community-aquired pneumonia from ages 6 to 18 years. The dependences of trace levels of zinc and copper in the serum of these children on the activity of the inflammatory process. Analysis of survey results

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service