Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33-0090

Sigma-Aldrich

L-Valine

JIS special grade, ≥99.0%

Synonym(s):

(S)-α-Aminoisovaleric acid, L-2-Amino-3-methylbutanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH(NH2)CO2H
CAS Number:
Molecular Weight:
117.15
Beilstein/REAXYS Number:
1721136
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
Pricing and availability is not currently available.

grade

JIS special grade

assay

≥99.0%

form

solid

availability

available only in Japan

concentration

11.8-12.1% N

mp

295-300 °C (subl.) (lit.)

SMILES string

CC(C)[C@H](N)C(O)=O

InChI

1S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)/t4-/m0/s1

InChI key

KZSNJWFQEVHDMF-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonia Jego et al.
Nature neuroscience, 16(11), 1637-1643 (2013-09-24)
Rapid-eye movement (REM) sleep correlates with neuronal activity in the brainstem, basal forebrain and lateral hypothalamus. Lateral hypothalamus melanin-concentrating hormone (MCH)-expressing neurons are active during sleep, but their effects on REM sleep remain unclear. Using optogenetic tools in newly generated
Felix Stickel et al.
Archives of internal medicine, 165(16), 1835-1840 (2005-09-15)
Oxidative stress plays an important pathogenic role in hereditary hemochromatosis (HHC) and chronic hepatitis C virus infection (CHC). Several enzymes involved in the degradation of reactive oxidants and xenobiotics, such as glutathione S-transferase P1 (GSTP1) and manganese superoxide dismutase (MnSOD)
Shankar Sachidhanandam et al.
Nature neuroscience, 16(11), 1671-1677 (2013-10-08)
Neocortical activity can evoke sensory percepts, but the cellular mechanisms remain poorly understood. We trained mice to detect single brief whisker stimuli and report perceived stimuli by licking to obtain a reward. Pharmacological inactivation and optogenetic stimulation demonstrated a causal
Ihab Daou et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(47), 18631-18640 (2013-11-22)
We report a novel model in which remote activation of peripheral nociceptive pathways in transgenic mice is achieved optogenetically, without any external noxious stimulus or injury. Taking advantage of a binary genetic approach, we selectively targeted Nav1.8(+) sensory neurons for
Adam J Mead et al.
Blood, 121(20), 4156-4165 (2013-03-29)
The association between somatic JAK2 mutation and myeloproliferative neoplasms (MPNs) is now well established. However, because JAK2 mutations are associated with heterogeneous clinical phenotypes and often occur as secondary genetic events, some aspects of JAK2 mutation biology remain to be

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service