Antibodies that impair Plasmodium falciparum merozoite invasion and intraerythrocytic development are one of several mechanisms that mediate naturally acquired immunity to malaria. Attempts to correlate anti-malaria antibodies with risk of infection and morbidity have yielded inconsistent results. Growth inhibition assays
Journal of cellular physiology, 228(11), 2222-2231 (2013-04-30)
FUsed in Sarcoma/Translocated in LipoSarcoma (FUS/TLS or FUS) has been linked to several biological processes involving DNA and RNA processing, and has been associated with multiple diseases, including myxoid liposarcoma and amyotrophic lateral sclerosis (ALS). ALS-associated mutations cause FUS to
The Journal of laboratory and clinical medicine, 131(5), 393-405 (1998-05-30)
D-Sorbitol (SOR) is safe, is easy to measure, and has an exceptionally high extraction ratio in the normal liver of 0.93+/-0.05 (mean+/-SD). Together with the general interest in hepatic hemodynamics, these facts motivated us to review the usefulness of this
Orphanet journal of rare diseases, 9, 199-199 (2014-12-19)
Charcot-Marie-Tooth type 1A disease (CMT1A) is a rare orphan inherited neuropathy caused by an autosomal dominant duplication of a gene encoding for the structural myelin protein PMP22, which induces abnormal Schwann cell differentiation and dysmyelination, eventually leading to axonal suffering
Cellular and molecular life sciences : CMLS, 71(21), 4275-4283 (2014-04-29)
Two-pore channel proteins (TPC) encode intracellular ion channels in both animals and plants. In mammalian cells, the two isoforms (TPC1 and TPC2) localize to the endo-lysosomal compartment, whereas the plant TPC1 protein is targeted to the membrane surrounding the large
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.