Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-3950

Sigma-Aldrich

Sodium propionate

SAJ first grade, ≥98.0%

Synonym(s):

Propionic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2COONa
CAS Number:
Molecular Weight:
96.06
Beilstein/REAXYS Number:
3566934
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥98.0%
grade:
SAJ first grade
form:
powder

grade

SAJ first grade

vapor pressure

<0.1 hPa ( 20 °C)

assay

≥98.0%

form

powder

availability

available only in Japan

mp

285-286 °C (lit.)

SMILES string

[Na+].CCC([O-])=O

InChI

1S/C3H6O2.Na/c1-2-3(4)5;/h2H2,1H3,(H,4,5);/q;+1/p-1

InChI key

JXKPEJDQGNYQSM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucio Zennaro et al.
Cancer cytopathology, 125(5), 341-348 (2017-02-01)
Cytopathology is a noninvasive and cost-effective method for detecting cancer cells in pleural effusions (PEs), although in many cases, the diagnostic performance is hindered by the paucity of significant cells or the lack of clear morphological criteria. This study presents
Aino-Liisa Alanne et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 74(7-8), 201-210 (2019-06-15)
Two edible, cultivable mushroom species of the family Strophariaceae, Kuehneromyces mutabilis (sheathed woodtuft) and Hypholoma capnoides (conifer tuft), were studied using proton nuclear magnetic resonance metabolomic approach. The variation in the metabolites of the two species and their metabolic behaviour
Virginia T Guidry et al.
Environmental health perspectives, 123(7), 705-711 (2015-03-18)
Exposures to environmental phenols and parabens may be harmful, especially in utero. Prior studies have demonstrated high within-person variability of urinary concentrations across pregnancy. We sought to measure phenol and paraben biomarker concentrations for the Norwegian Mother and Child Cohort
Licia Lama et al.
AMB Express, 4, 55-55 (2014-07-16)
Polysaccharidases from extremophiles are remarkable for specific action, resistance to different reaction conditions and other biotechnologically interesting features. In this article the action of crude extracts of thermophilic microorganisms (Thermotoga neapolitana, Geobacillus thermantarcticus and Thermoanaerobacterium thermostercoris) is studied using as
Bistra B Nankova et al.
PloS one, 9(8), e103740-e103740 (2014-08-30)
Alterations in gut microbiome composition have an emerging role in health and disease including brain function and behavior. Short chain fatty acids (SCFA) like propionic (PPA), and butyric acid (BA), which are present in diet and are fermentation products of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service