Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-3740

Sigma-Aldrich

Sodium phosphate dibasic dodecahydrate

99.0-100.5%, suitable for oxidant determination

Synonym(s):

sec-Sodium phosphate, Disodium hydrogen phosphate dodecahydrate, Disodium phosphate, di-Sodium hydrogen phosphate dodecahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4 · 12H2O
CAS Number:
Molecular Weight:
358.14
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
99.0-100.5%
grade:
SAJ special grade
form:
crystalline
Pricing and availability is not currently available.

grade

SAJ special grade

assay

99.0-100.5%

form

crystalline

availability

available only in Japan

pH

8.4-9.6 (25 °C, 35.8 g/L)

suitability

suitable for oxidant determination

SMILES string

O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].OP([O-])([O-])=O

InChI

1S/2Na.H3O4P.12H2O/c;;1-5(2,3)4;;;;;;;;;;;;/h;;(H3,1,2,3,4);12*1H2/q2*+1;;;;;;;;;;;;;/p-2

InChI key

DGLRDKLJZLEJCY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuichiro Yano et al.
Journal of the American College of Cardiology, 65(4), 327-335 (2015-01-31)
Isolated systolic hypertension (ISH), defined as systolic blood pressure (SBP) ≥140 mm Hg and diastolic blood pressure (DBP) <90 mm Hg, in younger and middle-aged adults is increasing in prevalence. The aim of this study was to assess the risk
Karen Sliwa et al.
Heart (British Cardiac Society), 100(24), 1967-1974 (2014-09-18)
Lack of evidence-based data on the spectrum of cardiovascular disease (CVD) in pregnancy or in the postpartum period, as well as on maternal and fetal outcome, provides challenges for treating physicians, particularly in areas of low resources. The objectives of
Matthew R Weir et al.
Journal of clinical hypertension (Greenwich, Conn.), 16(12), 875-882 (2014-10-21)
The effects of canagliflozin, a sodium glucose co-transporter 2 inhibitor, on blood pressure (BP) and osmotic diuresis- and intravascular volume reduction-related adverse events (AEs) were evaluated using pooled data from four placebo-controlled, phase 3 studies in patients with type 2
George G S Sandor et al.
International journal of cardiology, 179, 470-475 (2014-12-04)
Patients with Marfan (MFS) and Loeys-Dietz (LDS) syndromes have been shown to have abnormal aortic biophysical properties. The purpose of this study was to compare the effects of 12-months of therapy with atenolol or losartan on vascular function in young
Luciano Candilio et al.
Heart (British Cardiac Society), 101(3), 185-192 (2014-09-26)
Remote ischaemic preconditioning (RIPC), using brief cycles of limb ischaemia/reperfusion, is a non-invasive, low-cost intervention that may reduce perioperative myocardial injury (PMI) in patients undergoing cardiac surgery. We investigated whether RIPC can also improve short-term clinical outcomes. One hundred and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service