Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-3565

Sigma-Aldrich

Sodium oxalate solution

0.25 M

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOOCCOONa
CAS Number:
Molecular Weight:
134.00
Beilstein/REAXYS Number:
3631622
MDL number:
UNSPSC Code:
12352300
Pricing and availability is not currently available.

availability

available only in Japan

concentration

0.25 M
1/2 N

SMILES string

[Na+].[Na+].[O-]C(=O)C([O-])=O

InChI

1S/C2H2O4.2Na/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);;/q;2*+1/p-2

InChI key

ZNCPFRVNHGOPAG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiří Jan et al.
Water research, 47(2), 547-557 (2012-12-12)
Correct identification of P forms together with their main Fe and Al binding partners in non-calcareous sediments is of crucial importance for evaluation of P cycling in water bodies. In this paper, we assess extraction methods frequently used for this
Lu Gao et al.
International journal of phytoremediation, 15(4), 307-319 (2013-03-16)
Phytolacca americana L. can accumulate large amounts of heavy metals in its aerial tissues, especially cadmium (Cd) and manganese (Mn). It has great potential for use in phytoextraction of metals from multi-metal-contaminated soils. This study was conducted to further investigate
A Rosal et al.
Environmental technology, 33(16-18), 2149-2158 (2012-12-18)
Compost made from municipal solid waste (MSW) contains heavy metals that can interfere with the use of organic amendment in soil. In order to find effective ways to reduce the potential risk of heavy metals, we have investigated a novel
Liying Jiang et al.
Environmental technology, 34(1-4), 131-138 (2013-03-28)
Natural estrogens, such as 17beta-estradiol (E2), are the main substances responsible for estrogenic activity found in domestic sewage. In the work described herein, the degradation of E2 has been investigated by single ozonation and catalytic ozonation in the presence of
Lois Fernandez et al.
Clinical biochemistry, 46(4-5), 285-288 (2012-12-12)
There are no direct comparisons of blood glucose values in samples collected with barrier serum tubes (SST) and NaF/potassium oxalate (NaF/KOx) plasma tubes. Collection of samples in SST tubes can offer considerable savings and specimen processing advantages for national level

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service