All Photos(1)
About This Item
Linear Formula:
NaBrO3
CAS Number:
Molecular Weight:
150.89
MDL number:
UNSPSC Code:
12352300
grade
SAJ first grade
assay
≥99.0%
availability
available only in Japan
density
3.339 g/mL at 25 °C (lit.)
SMILES string
[Na+].[O-]Br(=O)=O
InChI
1S/BrHO3.Na/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1
InChI key
XUXNAKZDHHEHPC-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral - Eye Irrit. 2 - Ox. Sol. 1 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
5.1A - Strongly oxidizing hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Shu-Ichi Shinohara et al.
Chemical communications (Cambridge, England), (39)(39), 4735-4737 (2008-10-03)
We demonstrate the chemical and optical control of the self-sustaining peristaltic motion of a structural colored porous hydrogel.
Wagner A Alves et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 58(7), 1395-1399 (2002-06-27)
The splitting of v1 (794 cm(-1)) and v3 (805 cm(-1)) modes in the stretching region of the bromate (C3v) was observed for the first time in dimethyl sulfoxide (DMSO) using vibrational spectroscopy. Depolarization measurements allowed to assign the asymmetric and
Diana I Roncaglia et al.
Physical chemistry chemical physics : PCCP, 13(16), 7426-7432 (2011-03-25)
The Belousov-Zhabotinsky reaction was studied under the influence of nanometric confinements induced by a complex polymer, the PAMAM-G4 dendrimers. They are well-defined in both molecular weight and architecture and are capable of molecular inclusion, making "unimolecular active micelles". The effect
T Umemura et al.
Journal of cancer research and clinical oncology, 119(8), 463-469 (1993-01-01)
Accumulation of alpha 2u-globulin and induction of cell proliferation were examined in kidneys of rats exposed to KBrO3, KBr or NaBrO3 in their drinking water. Hyaline droplets observed after KBrO3 or NaBrO3 administration to male rats were specifically immunostained for
Cristobal Viedma
Astrobiology, 7(2), 312-319 (2007-05-08)
Chiral symmetry breaking occurs when a physical or chemical process spontaneously generates a large excess of one of the two enantiomers--left-handed (L) or right-handed (D)--with no preference as to which of the two enantiomers is produced. From the viewpoint of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service