Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-1390

Sigma-Aldrich

Silver(I) oxide

SAJ special grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ag2O
CAS Number:
Molecular Weight:
231.74
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥99.0%
grade:
SAJ special grade
form:
powder
Pricing and availability is not currently available.

grade

SAJ special grade

assay

≥99.0%

form

powder

availability

available only in Japan

storage temp.

15-25°C

SMILES string

[Ag]O[Ag]

InChI

1S/2Ag.O

InChI key

KHJDQHIZCZTCAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 1

Storage Class

5.1A - Strongly oxidizing hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weijia Zhou et al.
Physical chemistry chemical physics : PCCP, 12(45), 15119-15123 (2010-10-22)
Electrostatic self-assembled Ag(2)O/TiO(2) nanobelts heterostructure was synthesized via simple physical mixing of Ag(2)O nanoparticles and TiO(2) nanobelts. The morphologies and microstructures of Ag(2)O/TiO(2) nanobelt heterostructure were characterized by high resolution transmission electron microscopy. The interface dominated high UV photocatalytic activity
Chiara Uboldi et al.
Mutation research, 745(1-2), 11-20 (2011-11-19)
Although amorphous silica nanoparticles (aSiO(2)NPs) are believed to be non-toxic and are currently used in several industrial and biomedical applications including cosmetics, food additives and drug delivery systems, there is still no conclusive information on their cytotoxic, genotoxic and carcinogenic
Arixin Bo et al.
Journal of hazardous materials, 246-247, 199-205 (2013-01-15)
Emergency treatment of radioactive material leakage and safety disposal of nuclear waste is a constant concern all along with the development of radioactive materials applications. To provide a solution, titanate with large surface area (143 m(2)g(-1)) and a lamina morphology
Carolyn P Ojano-Dirain et al.
International journal of pediatric otorhinolaryngology, 77(2), 223-227 (2012-12-04)
Tympanostomy tube (TT) surface modifications have been promoted as a means of reducing biofilm formation, otorrhea, and occlusion. The goal of this study was to determine if biofilm formation on silicone TTs could be prevented by commercially available surface coatings.
G Belcaro et al.
Panminerva medica, 53(3 Suppl 1), 29-33 (2011-12-14)
Venous ulcers are a common, chronic medical and social problem. These ulcers are difficult to heal in most patients with sustained venous hypertension. This pilot product evaluation registry study has evaluated the efficacy and safety of an antimicrobial silver oxide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service