Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-5640

Sigma-Aldrich

Potassium sulfite

SAJ first grade, ≥95.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2SO3
CAS Number:
Molecular Weight:
158.26
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
SAJ first grade
assay:
≥95.0%
Pricing and availability is not currently available.

grade

SAJ first grade

assay

≥95.0%

availability

available only in Japan

dilution

(for analytical testing)

SMILES string

[K+].[K+].[O-]S([O-])=O

InChI

1S/2K.H2O3S/c;;1-4(2)3/h;;(H2,1,2,3)/q2*+1;/p-2

InChI key

BHZRJJOHZFYXTO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinjian Shi et al.
Nature communications, 5, 4775-4775 (2014-09-03)
Tungsten trioxide/bismuth vanadate heterojunction is one of the best pairs for solar water splitting, but its photocurrent densities are insufficient. Here we investigate the advantages of using helical nanostructures in photoelectrochemical solar water splitting. A helical tungsten trioxide array is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service