Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-5090

Sigma-Aldrich

Potassium perchlorate

SAJ special grade, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KClO4
CAS Number:
Molecular Weight:
138.55
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥99.5%
grade:
SAJ special grade
form:
crystalline
Pricing and availability is not currently available.

grade

SAJ special grade

assay

≥99.5%

form

crystalline

reaction suitability

reagent type: oxidant

availability

available only in Japan

pH

5.0-6.5 (25 °C, 13.9 g/L)

mp

400 °C (dec.) (lit.)

SMILES string

[K+].[O-]Cl(=O)(=O)=O

InChI

1S/ClHO4.K/c2-1(3,4)5;/h(H,2,3,4,5);/q;+1/p-1

InChI key

YLMGFJXSLBMXHK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

pictograms

Flame over circleExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Ox. Sol. 1

Storage Class

5.1A - Strongly oxidizing hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manuel E Ortiz-Santaliestra et al.
Archives of environmental contamination and toxicology, 53(4), 639-646 (2007-07-28)
Amphibians are sensitive to a great variety of agrochemicals. Nitrate compounds are commonly applied as fertilizers, whereas perchlorate salts occur as by-products of the military industry and in some nitrate fertilizers. Both compounds are highly soluble and can easily diffuse
V Jahagirdar et al.
Journal of neuroendocrinology, 24(8), 1126-1134 (2012-03-23)
Steroid hormones exert profound effects on the development of brain areas controlling complex cognitive function in adulthood. One class, progestins, may contribute by acting on the progestin receptor (PR), which is transiently expressed in a critical layer of developing cortex:
Caterina Constantinou et al.
Journal of proteome research, 10(2), 869-879 (2010-10-30)
Although adult-onset hypothyroidism (AOH) has been connected to neural activity alterations, including movement, behavioral, and mental dysfunctions, the underlying changes in brain metabolic physiology have not been investigated in a systemic and systematic way. The current knowledge remains fragmented, referring
Florian Schmidt et al.
Aquatic toxicology (Amsterdam, Netherlands), 109, 47-58 (2011-12-30)
The increasing pollution of aquatic habitats with anthropogenic compounds has led to various test strategies to detect hazardous chemicals. However, information on effects of pollutants in the thyroid system in fish, which is essential for growth, development and parts of
Baohua Gu et al.
Environmental science & technology, 41(17), 6277-6282 (2007-10-17)
Treatment of perchlorate-contaminated water using highly selective, regenerable ion-exchange and perchlorate-destruction technologies was demonstrated at a field site in California. Four treatment and four regeneration cycles were carried out, and no significant deterioration of resin performance was noted in 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service