Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-4610

Sigma-Aldrich

Potassium fluoride

SAJ special grade, ≥98.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KF
CAS Number:
Molecular Weight:
58.10
Beilstein/REAXYS Number:
3902818
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥98.5%
grade:
SAJ special grade
form:
crystalline
Pricing and availability is not currently available.

grade

SAJ special grade

assay

≥98.5%

form

crystalline

availability

available only in Japan

mp

858 °C (lit.)

SMILES string

[F-].[K+]

InChI

1S/FH.K/h1H;/q;+1/p-1

InChI key

NROKBHXJSPEDAR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

A versatile fluoride ion source used in a study of ion-specific swelling and de-swelling of ampholytic polymer gels, in the room temperature oxidation of noble metals in HF, and in the measurement of electronic polarizabilities of ions in polymers of alkali halides.

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marianna Mamusa et al.
Faraday discussions, 181, 193-209 (2015-05-12)
Maghemite (γ-Fe2O3) nanoparticles (NPs) can be successfully dispersed in a protic ionic liquid, ethylammonium nitrate (EAN), by transfer from aqueous dispersions into EAN. As the aqueous systems are well controlled, several parameters can be tuned. Their crucial role towards the
Mónica Calvo-Polanco et al.
Physiologia plantarum, 135(1), 51-61 (2009-01-06)
Black spruce (Picea mariana), white spruce (Picea glauca) and jack pine (Pinus banksiana) were inoculated with Suillus tomentosus and subjected to potassium fluoride (1 mM KF and 5 mM KF) in the presence and absence of 60 mM NaCl. The
E Gershonov et al.
The Journal of organic chemistry, 74(1), 329-338 (2008-12-05)
A facile solvent-free hydrolysis (chemical destruction) of the warfare agents VX (O-ethyl S-2-(diisopropylamino)ethyl methylphosphonothioate), GB (O-isopropyl methylphosphonofluoridate or sarin), and HD (2,2'-dichloroethyl sulfide or sulfur mustard) upon reaction with various solid-supported fluoride reagents is described. These solid reagents include different
Mingming Fan et al.
Bioresource technology, 104, 447-450 (2011-12-16)
Transesterification of soybean oil with methanol was carried out in the presence of CaO-MgO and KF-modified CaO-MgO catalysts at atmospheric pressure. While the methyl ester yield for the CaO-MgO catalyst with a ratio of 8:2 (CaO:MgO) was 63.6%, it was
Immanuel Kalcher et al.
The Journal of chemical physics, 130(13), 134507-134507 (2009-04-10)
The structure of aqueous LiCl, NaCl, KCl, CsCl, KF, and NaI solutions is calculated by molecular dynamics (MD) simulations of the frequently employed Dang force-field in SPC/E water. By using liquid state theory, we integrate the structure to obtain the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service