Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-3890

Sigma-Aldrich

Potassium

SAJ first grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
K
CAS Number:
Molecular Weight:
39.10
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

SAJ first grade

vapor pressure

0.09 mmHg ( 260 °C)

reaction suitability

reagent type: reductant

availability

available only in Japan

resistivity

6.1 μΩ-cm, 20°C

bp

760 °C (lit.)

mp

64 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

SMILES string

[K]

InChI

1S/K

InChI key

ZLMJMSJWJFRBEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Combines under pressure with silver metal to form 2 new compounds: K2Ag and K3Ag.†

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - Water-react 1

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicole Schmitt et al.
Physiological reviews, 94(2), 609-653 (2014-04-03)
About 10 distinct potassium channels in the heart are involved in shaping the action potential. Some of the K+ channels are primarily responsible for early repolarization, whereas others drive late repolarization and still others are open throughout the cardiac cycle.
Glucose-insulin-potassium revived: current status in acute coronary syndromes and the energy-depleted heart.
Alexandra N Grossman et al.
Circulation, 127(9), 1040-1048 (2013-03-06)
Jordan E Bisanz et al.
Applied and environmental microbiology, 81(15), 4965-4975 (2015-05-17)
The nutritional status of pregnant women is vital for healthy outcomes and is a concern for a large proportion of the world's population. The role of the microbiota in pregnancy and nutrition is a promising new area of study with
Stefan D Anker et al.
European journal of heart failure, 17(10), 1050-1056 (2015-05-27)
Hyperkalaemia in heart failure patients limits use of cardioprotective renin-angiotensin-aldosterone system inhibitors (RAASi). Sodium zirconium cyclosilicate (ZS-9) is a selective potassium ion trap, whose mechanism of action may allow for potassium binding in the upper gastrointestinal tract as early as
David K Packham et al.
The New England journal of medicine, 372(3), 222-231 (2014-11-22)
Hyperkalemia (serum potassium level, >5.0 mmol per liter) is associated with increased mortality among patients with heart failure, chronic kidney disease, or diabetes. We investigated whether sodium zirconium cyclosilicate (ZS-9), a novel selective cation exchanger, could lower serum potassium levels

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service