Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-2670

Sigma-Aldrich

Phosphorus, red

CP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
P
CAS Number:
Molecular Weight:
30.97
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

CP

vapor density

0.02 (vs air)

vapor pressure

0.03 mmHg ( 21 °C)
1 mmHg ( 76.6 °C)

form

powder

autoignition temp.

500 °F

reaction suitability

reagent type: ligand

availability

available only in Japan

resistivity

10 μΩ-cm, 20°C

pH

~3 (37 °C, 10 g/L)

mp

416 °C (subl.) (lit.)

density

2.34 g/mL at 25 °C (lit.)

functional group

phosphine

SMILES string

[P]

InChI

1S/P

InChI key

OAICVXFJPJFONN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 3 - Flam. Sol. 2

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manuel Delgado-Baquerizo et al.
Nature, 502(7473), 672-676 (2013-11-01)
The biogeochemical cycles of carbon (C), nitrogen (N) and phosphorus (P) are interlinked by primary production, respiration and decomposition in terrestrial ecosystems. It has been suggested that the C, N and P cycles could become uncoupled under rapid climate change
Bon-Chul Koo et al.
Science (New York, N.Y.), 342(6164), 1346-1348 (2013-12-18)
Phosphorus ((31)P), which is essential for life, is thought to be synthesized in massive stars and dispersed into interstellar space when these stars explode as supernovae (SNe). Here, we report on near-infrared spectroscopic observations of the young SN remnant Cassiopeia
Jacques C Finlay et al.
Science (New York, N.Y.), 342(6155), 247-250 (2013-10-12)
Human activities have increased the availability of reactive nitrogen in many ecosystems, leading to negative impacts on human health, biodiversity, and water quality. Freshwater ecosystems, including lakes, streams, and wetlands, are a large global sink for reactive nitrogen, but factors
Yuki Fujita et al.
Nature, 505(7481), 82-86 (2013-11-19)
Plant species diversity in Eurasian wetlands and grasslands depends not only on productivity but also on the relative availability of nutrients, particularly of nitrogen and phosphorus. Here we show that the impacts of nitrogen:phosphorus stoichiometry on plant species richness can
Zhiguo Cao et al.
Environment international, 65, 100-106 (2014-02-01)
This study documents the temporal variability in concentrations of flame retardants (FRs) in floor dust from three offices in Beijing, China. Dust from Office A (OAD) was collected weekly from March to August, 2012, and sampling of dust from Office

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service