Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-2495

Sigma-Aldrich

Phosphate standard solution

0.1 mg/mL PO43-

Synonym(s):

Potassium Phosphate Monobasic solution, Monopotassium phosphate, Potassium dihydrogen phosphate solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
Pricing and availability is not currently available.

availability

available only in Japan

concentration

0.1 mg/mL PO43-

SMILES string

[K+].OP(O)([O-])=O

InChI

1S/K.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

GNSKLFRGEWLPPA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jayasankar V Valiyaparambil et al.
The International journal of oral & maxillofacial implants, 27(5), 1271-1277 (2012-10-12)
To examine the relationship between dental cone beam computed tomography (CBCT) gray scale values and Hounsfield units (HU), and whether the gray values of edentulous sites correlate with the subjective clinical bone quality assessed at surgery. Two radiographic phantoms containing
Pingzhong Feng et al.
Journal of bioscience and bioengineering, 114(4), 405-410 (2012-06-16)
To evaluate the potential of the green agla Chlorella zofingiensis as a feedstock for biodiesel production, the effects of nitrogen and phosphate on lipid accumulation and growth of C. zofingiensis were studied. The maximum specific growth rate (μ(max)) reached 2.15
R Krishnamurthy et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 104, 310-314 (2013-01-01)
Potassium Dihydrogen Phosphate (KDP) doped with L-aspartic acid has been grown by solvent slow evaporation technique from a mixture of aqueous solution of KDP and 0.7% of L-aspartic acid at room temperature. The grown crystals were characterized by powder X-ray
Kamran Ajmal Syed et al.
The Journal of chemical physics, 138(2), 024901-024901 (2013-01-17)
The efflorescence of an individual KH(2)PO(4) droplet on Teflon substrate was investigated by micro-Raman spectroscopy. With the decrease of relative humidity (RH) from 98.0% to 73.0%, the KH(2)PO(4) droplet lost water gradually and entered into supersaturated state, which was reflected
Marko Usaj et al.
The Journal of membrane biology, 245(9), 583-590 (2012-07-31)
The fusogenic state of the cell membrane can be induced by external electric field. When two fusogenic membranes are in close contact, cell fusion takes place. An appropriate hypotonic treatment of cells before the application of electric pulses significantly improves

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service