Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21-2600

Sigma-Aldrich

Nitromethane

SAJ first grade, ≥90.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH3NO2
CAS Number:
Molecular Weight:
61.04
Beilstein/REAXYS Number:
1698205
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:
grade:
SAJ first grade
assay:
≥90.0%
bp:
101.2 °C (lit.)
vapor pressure:
2.7 mmHg

grade

SAJ first grade

vapor density

2.1 (vs air)

vapor pressure

2.7 mmHg

assay

≥90.0%

form

liquid

autoignition temp.

784 °F

expl. lim.

7.3 %, 33 °F

availability

available only in Japan

dilution

(for analytical testing)

refractive index

n20/D 1.382 (lit.)

pH

6.4 (20 °C, 0.01 g/L)

bp

101.2 °C (lit.)

mp

−29 °C (lit.)

density

1.127 g/mL at 25 °C (lit.)

SMILES string

C[N+]([O-])=O

InChI

1S/CH3NO2/c1-2(3)4/h1H3

InChI key

LYGJENNIWJXYER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 3 - Repr. 2

Storage Class

4.1A - Other explosive hazardous materials

wgk_germany

WGK 2

flash_point_f

95.0 °F

flash_point_c

35 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Szekely et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 959, 1-4 (2014-04-22)
Since 2008 a significant amount of work has focused on the development of methods to analyze contaminated heparin. This work focuses on utilizing heparin's ability to serve as a chiral selector as a means for determining contamination. Specifically, the effect
Huachang Hong et al.
The Science of the total environment, 444, 196-204 (2012-12-29)
The formations of THMs, HAAs, and HNMs from chlorination and chloramination of water from Jinlan Reservoir were investigated in this study. Results showed that monochloramine rather than chlorine generally resulted in lower concentration of DBPs, and the DBPs formation varied
Naomi Rom et al.
The journal of physical chemistry. A, 115(36), 10181-10202 (2011-08-05)
The decomposition mechanism of hot liquid nitromethane at various compressions was studied using reactive force field (ReaxFF) molecular dynamics simulations. A competition between two different initial thermal decomposition schemes is observed, depending on compression. At low densities, unimolecular C-N bond
Xin Yang et al.
Environmental science & technology, 46(23), 12832-12838 (2012-11-17)
Nitrogenous disinfection byproducts (N-DBPs) such as haloacetonitriles (HANs) and halonitromethanes (HNMs) are formed during water chlorination. Preozonation is sometimes applied to control trihalomethane (THM) formation, but this may risk promoting the formation of HNMs and HANs. The role of ozone
Junhong Shan et al.
Chemosphere, 86(4), 323-328 (2011-11-01)
In this study, the effects of pH, bromide and nitrite on the formation of halonitromethanes (HNMs) and trihalomethanes (THMs) from eight amino acids (glycine, alanine, serine, cysteine, aspartic acid, glutamic acid, lysine and histidine) and four amino sugars (glucosamine, galactosamine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service