Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07-6125

Sigma-Aldrich

DL-Dithiothreitol

SAJ special grade, ≥98.0%

Synonym(s):

threo-1,4-Dimercapto-2,3-butanediol, Cleland’s reagent, DTT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(OH)CH(OH)CH2SH
CAS Number:
Molecular Weight:
154.25
Beilstein/REAXYS Number:
1719757
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ special grade

assay

≥98.0%

availability

available only in Japan

mp

41-44 °C (lit.)

SMILES string

O[C@H](CS)[C@H](O)CS

InChI

1S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8H,1-2H2/t3-,4-/m1/s1

InChI key

VHJLVAABSRFDPM-QWWZWVQMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

An excellent reagent for maintaining SH groups in reduced state; quantitatively reduces disulfides. DTT is effective in sample buffers for reducing protein disulfide bonds prior to SDS-PAGE. DTT can also be used for reducing the disulfide bridge of the cross-linker N,N′-bis(acryloyl)cystamine to break apart the matrix of a polyacrylamide gel. DTT is less pungent and is less toxic than 2-mercaptoethanol. Typically, a seven fold lower concentration of DTT (100 mM) is needed than is used for 2-mercaptoethanol (5% v/v, 700 mM).

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin-Gu Lee et al.
Nature communications, 4, 1568-1568 (2013-03-07)
In eukaryotes, deubiquitinases (DUBs) remove ubiquitin conjugates from diverse substrates, altering their stabilities, localizations or activities. Here we show that many DUBs of the USP and UCH subfamilies can be reversibly inactivated upon oxidation by reactive oxygen species in vitro
Xueqian Liu et al.
The Journal of biological chemistry, 288(10), 7230-7240 (2013-01-25)
In Pichia pastoris, the peroxisomal targeting signal 2 (PTS2)-dependent peroxisomal matrix protein import pathway requires the receptor, Pex7, and its co-receptor Pex20. A conserved lysine (Lys(19)) near the N terminus of Pex20 is required for its polyubiquitination and proteasomal degradation
František Štork et al.
Environmental science and pollution research international, 20(8), 5502-5511 (2013-02-26)
Effect of nitric oxide donor (sodium nitroprusside, SNP, 500 μM) or hydrogen peroxide scavenger (dithiothreitol, DTT, 500 μM) on cadmium (Cd) or copper (Cu) uptake (150 μM solutions) and toxicity using Scenedesmus quadricauda was studied. Combined treatments (Cd or Cu
Hanayo Ueoka-Nakanishi et al.
The FEBS journal, 280(14), 3220-3231 (2013-04-26)
Thioredoxin (Trx) is a key player in redox homeostasis in various cells, modulating the functions of target proteins by catalyzing a thiol-disulfide exchange reaction. Target proteins of cytosolic Trx-h of higher plants were studied, particularly in the plasma membrane, because
Elaine C Lima de Souza et al.
Endocrinology, 154(5), 1948-1955 (2013-04-03)
The thyroid hormone (TH) transporter monocarboxylate transporter 8 (MCT8) is crucial for brain development as demonstrated by the severe psychomotor retardation in patients with MCT8 mutations. MCT8 contains 10 residues of the reactive amino acid cysteine (Cys) whose functional roles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service