Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05-6030

Sigma-Aldrich

Copper(II) nitrate trihydrate

SAJ first grade, 77.0-80.0% anhydrous basis

Synonym(s):

Cupric nitrate trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cu(NO3)2 · 3H2O
CAS Number:
Molecular Weight:
241.60
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ first grade

assay

77.0-80.0% anhydrous basis

form

solid

availability

available only in Japan

storage temp.

15-25°C

SMILES string

O.O.O.[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Cu.2NO3.3H2O/c;2*2-1(3)4;;;/h;;;3*1H2/q+2;2*-1;;;

InChI key

SXTLQDJHRPXDSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Imtiyaz Yousuf et al.
Dalton transactions (Cambridge, England : 2003), 44(22), 10330-10342 (2015-05-15)
A new chromone-appended Cu(ii) drug entity () was designed and synthesized as a potential anticancer chemotherapeutic agent. The structural elucidation was carried out thoroughly by elemental analysis, FT-IR, EPR, ESI-MS and single crystal X-ray crystallography. Complex resulted from the in
Adetayo M Mustapha et al.
Journal of mass spectrometry : JMS, 50(1), 145-152 (2015-01-21)
Efforts to improve agricultural productivity have led to a growing dependency on organophosphorus pesticides. Phosphorothioate and phosphorodithioate pesticides are organophosphorus pesticide subclasses with widespread application for the control of insects feeding on vegetables and fruits. However, even low doses of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service