Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05-1540

Sigma-Aldrich

Carbon disulfide

JIS special grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CS2
CAS Number:
Molecular Weight:
76.14
Beilstein/REAXYS Number:
1098293
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

JIS special grade

vapor density

2.67 (vs air)

vapor pressure

5.83 psi

assay

≥99.0%

form

liquid

autoignition temp.

212 °F

expl. lim.

50 %

availability

available only in Japan

refractive index

n20/D 1.627 (lit.)

bp

46 °C (lit.)

mp

−112-−111 °C (lit.)

density

1.266 g/mL at 25 °C (lit.)

SMILES string

S=C=S

InChI

1S/CS2/c2-1-3

InChI key

QGJOPFRUJISHPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

-22.0 °F

flash_point_c

-30 °C

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Kilaru et al.
Fungal genetics and biology : FG & B, 79, 125-131 (2015-06-21)
Fluorescent proteins (FPs) are powerful tools to investigate intracellular dynamics and protein localization. Cytoplasmic expression of FPs in fungal pathogens allows greater insight into invasion strategies and the host-pathogen interaction. Detection of their fluorescent signal depends on the right combination
M Schuster et al.
Fungal genetics and biology : FG & B, 79, 132-140 (2015-06-21)
The use of fluorescent proteins (FPs) in plant pathogenic fungi provides valuable insight into their intracellular dynamics, cell organization and invasion mechanisms. Compared with green-fluorescent proteins, their red-fluorescent "cousins" show generally lower fluorescent signal intensity and increased photo-bleaching. However, the
Mark B van Eldijk et al.
Chemical communications (Cambridge, England), 49(71), 7770-7772 (2013-06-19)
CS2 hydrolase, a zinc-dependent enzyme that converts carbon disulfide to carbon dioxide and hydrogen sulfide, exists as a mixture of octameric ring and hexadecameric catenane forms in solution. A combination of size exclusion chromatography, multi-angle laser light scattering, and mass
Huan Liu et al.
Journal of hazardous materials, 235-236, 298-306 (2012-08-21)
Chemical conditioners are often used to enhance sewage sludge dewaterability through altering sludge properties and flocs structure, both affect odorous compounds emissions not only during sludge conditioning but also in subsequent sludge disposal. This study was to investigate emission characteristics
Continuous direct spinning of fibers of single-walled carbon nanotubes with metallic chirality.
Rajyashree M Sundaram et al.
Advanced materials (Deerfield Beach, Fla.), 23(43), 5064-5068 (2011-10-11)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service