Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05-1100

Sigma-Aldrich

Calcium sulfate dihydrate

SAJ first grade, ≥95.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaSO4 · 2H2O
CAS Number:
Molecular Weight:
172.17
EC Number:
MDL number:
UNSPSC Code:
12352300
Pricing and availability is not currently available.

grade

SAJ first grade

assay

≥95.0%

availability

available only in Japan

SMILES string

O.O.[Ca++].[O-]S([O-])(=O)=O

InChI

1S/Ca.H2O4S.2H2O/c;1-5(2,3)4;;/h;(H2,1,2,3,4);2*1H2/q+2;;;/p-2

InChI key

PASHVRUKOFIRIK-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sung-Yen Lin et al.
The Journal of hand surgery, 38(1), 104-109 (2012-12-04)
Enchondroma of the hand with a pathologic fracture is generally treated by tumor curettage and bone grafting after the fracture has healed. However, delayed surgery postpones definitive diagnosis and prolongs the period of disability. We have treated pathologic fractures in
Eun Young Rha et al.
Plastic and reconstructive surgery, 133(1), 14-20 (2014-01-01)
The authors have developed a simple and intuitive method for measuring breast volume based on three-dimensional simulated images of magnetic resonance imaging scans to accurately estimate breast volume before breast reconstruction. The authors performed a retrospective review of the medical
Aletta Hazeveld et al.
American journal of orthodontics and dentofacial orthopedics : official publication of the American Association of Orthodontists, its constituent societies, and the American Board of Orthodontics, 145(1), 108-115 (2014-01-01)
Rapid prototyping is a fast-developing technique that might play a significant role in the eventual replacement of plaster dental models. The aim of this study was to investigate the accuracy and reproducibility of physical dental models reconstructed from digital data
Andrew G Park et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 31(5), 776-782 (2012-12-15)
A significant number of lower extremity fractures result in mal-union necessitating effective treatments to restore ambulation. Prior research in diabetic animal fracture models demonstrated improved healing following local insulin application to the fracture site and indicated that local insulin therapy
Barbara Morasch
Environmental pollution (Barking, Essex : 1987), 173, 133-137 (2012-12-04)
Karst systems represent important yet vulnerable drinking water resources. A wide spectrum of pollutants may be released into karst groundwater from agriculture, livestock farming, private households, and industry. This work provides an overview on the occurrence and dynamics of micropollutants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service