Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05-0950

Sigma-Aldrich

Calcium phosphate monobasic monohydrate

SAJ special grade, ≥90.0%

Synonym(s):

Calcium bis(dihydrogenphosphate) monohydrate, Calcium dihydrogenphosphate monohydrate, Primary calcium phosphate monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(H2PO4)2 · H2O
CAS Number:
Molecular Weight:
252.07
EC Number:
MDL number:
UNSPSC Code:
12352300
Pricing and availability is not currently available.

grade

SAJ special grade

assay

≥90.0%

availability

available only in Japan

SMILES string

O.[Ca++].OP(O)([O-])=O.OP(O)([O-])=O

InChI

1S/Ca.2H3O4P.H2O/c;2*1-5(2,3)4;/h;2*(H3,1,2,3,4);1H2/q+2;;;/p-2

Inchi Key

ZBZJARSYCHAEND-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dan Meng et al.
Materials science & engineering. C, Materials for biological applications, 47, 266-272 (2014-12-11)
Calcium phosphate cements (CPCs) have been widely used as bone graft substitutes. However, the undesirable osteoinductivity and slow degradability of CPCs greatly hamper their clinical application. The aim of this study was to synthesize a type of injectable, bioactive cement.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service