Recommended Products
usage
sufficient for 96 multiwell tests
manufacturer/tradename
Roche
shipped in
wet ice
storage temp.
2-8°C
General description
Specificity
Application
Packaging
Preparation Note
Working solution: Preparation of the solutions
- Use the contents of bottle 1 undiluted at 15 to 25 °C before use.
- Place three tablets from bottle 2 in a beaker and dissolve in 1 ml of buffer from bottle 1; use the tweezers provided to remove the tablets from the bottle. The resulting solution is reaction mixture 2 and is sufficient for about 20 incubation assays.
- Dissolve the contents of one bottle 3 in 0.6 ml of double-dist. water to give solution 3.
- Use the contents of bottle 4 undiluted.
- Use the contents of bottle 5 undiluted.
- Dilute the contents
Storage conditions (working solution): Reaction mixture 2: Prepare immediately before use and bring to 15 to 25 °C before use
Color reagent I: sulfanilamide, stabilized, 8 ml
Potassium nitrate standard, aliquots: stable at -15 to -25 °C for 3 month
Other Notes
Kit Components Only
- Potassium Phosphate Buffer, pH 7.5
- Coenzyme
- Nitrate Reductase (2)
- Color Reagent I, consisting of sulfanilamide
- Color Reagent II, consisting of N-(1-naphthyl)ethylendiamine dihydrochloride
- Potassium nitrate standard 80.0 mM
- Microtiter plate, a frame with 12 modules 8 wells each
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Resp. Sens. 1 - Skin Sens. 1
Storage Class
8B - Non-combustible corrosive hazardous materials
wgk_germany
WGK 2
flash_point_f
does not flash
flash_point_c
does not flash
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service