Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11754599001

Roche

Nonidet P40 Substitute

Synonym(s):

nonidet, nonidet p40

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

solution

packaging

pkg of 100 mL

manufacturer/tradename

Roche

absorption

= 0.29 at 254 nm at 0.05 % (w/v)

Application

Nonidet P40 is:
  • Used for solubilizing membrane proteins during isolation of membrane-protein complexes
  • Included in buffers for microplate assays
  • Included in buffers for microtiterplate assays
  • Used to stabilize enzymes such as Taq DNA polymerase

Preparation Note

Working concentration: 0.1 to 1%
Note: Use these values as a starting point. The optimal working concentration can differ depending on the application.
Storage conditions (working solution): Store the solution under nitrogen at 2 to 8 °C.

Other Notes

For life science research only. Not for use in diagnostic procedures.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

closed cup

flash_point_c

closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service