Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11376454001

Roche

Epidermal Growth Factor, human (hEGF)

recombinant (E. coli)

Synonym(s):

EGF, human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

biological source

human

recombinant

expressed in E. coli

assay

>95% (HPLC)

form

lyophilized

potency

<0.5 ng/mL EC50

mol wt

6200 Da

packaging

pkg of 100 μg

manufacturer/tradename

Roche

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

color

white

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... EGF(1950)

General description

Recombinant Epidermal Growth Factor, human (hEGF), is produced in E. coli and purified by standard chromatographic techniques. It is a 100μg white lyophilizate, filtered through 0.2μm pore size membrane, recombinant from E. coli.
Epidermal growth factor (EGF) is a small mitogenic polypeptide which is present in many mammalian species and is distributed throughout a wide number of tissues and body fluids EGF is produced in the tubular cells of the submaxillary gland of the mouse, in the acinar cells of the human submaxillary gland, and in the human duodenal glands.[1][2]
Human EGF is identical to β-urogastrone, a polypeptide which was recognized and isolated on the basis of its ability to inhibit gastric acid secretion.[1][2][3] 37 amino acids of the 53 amino acids comprising the longer urogastrone (human EGF) and mouse EGF are common to both peptides (70% homology) and the three disulfide bonds are formed in the same relative positions. The biological activity of human EGF is similar to that of mouse EGF.[4] The cellular receptor for EGF is the best understood growth factor receptor. The oncogene v-erbB codes for a product homologous to a portion of the EGF receptor in which the EGF-binding domain has been deleted. Evidence exists suggesting that this truncation of the EGF receptor may lead to constitutive activation without requirement for ligand binding.[5]
EGF stimulates the proliferation and differentiation of awide variety of cells of ectodermal and mesodermal origin (e.g., fibroblasts, glial cells, keratinocytes, epithelial cells, endothelial cells, chondrocytes).[1][2][5] EGF is a constituent of many serum-free media formulations for a variety of cells.

Specificity

Species Specificity: Human EGF is effective on human and mouse cells.

Specific activity: <0.5 ng/ml, at least the same specific activity (EC50) compared to the indicated standard is guaranteed.

Application

Epidermal growth factor (EGF) stimulates the proliferation and differentiation of a wide variety of cells of ectodermal and mesodermal origin, and is a constituent of many serum-free media formulations. It has been used in the cell differentiation assay.[6]

Sequence

Chain Length 54 AA
The primary structure of recombinant, human EGF (one polypeptide chain, 54 amino acids) is identical to that of human, natural EGF (β-urogastrone) (53 amino acids), however, recombinant EGF has an additional methionine at the amino-terminus.

Unit Definition

Unit Conversion: 1 BM unit = 3.25 NBSB units (natural IL-2)1 BM unit = 3.25 NBSB units (natural IL-2)

Unit Definition: EC50 defintion: The concentration of hEGF that is required to support half-maximal stimulation of cell proliferation (MTT cleavage) with MK cells.

Physical form

White lyophilizate, filtered through 0.2 μm pore size membrane

Preparation Note

Working concentration: 0.5 to 20 ng/ml
Human EGF exerts its biological activity in the concentration range of 0.5 to 20 ng/ml. Recommended concentration for serum-free cell culture is 1 to 10 ng/ml
Working solution: In sterile water (final concentration: 500 μg/ml), further dilution with medium or PBS (phosphate buffered saline) containing 1 mg/ml BSA (bovine serum albumin), or 1 to 10% serum.
Storage conditions (working solution): -15 to -25 °C
It is recommended to store the reconstituted solution in aliquots at -15 to -25 °C.
Note: Avoid repeated freezing and thawing.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Epidermal growth factor.
Stanley C
In Vitro Cellular & Developmental Biology, 23, 239-246 (1987)
David T Asuzu et al.
Cell reports, 40(8), 111223-111223 (2022-08-25)
Sporadic pituitary adenomas occur in over 10% of the population. Hormone-secreting adenomas, including those causing Cushing's disease (CD), cause severe morbidity and early mortality. Mechanistic studies of CD are hindered by a lack of in vitro models and control normal human
Epidermal growth factor.
G Carpenter et al.
Annual review of biochemistry, 48, 193-216 (1979-01-01)
Isolation and structure of urogastrone and its relationship to epidermal growth factor.
H Gregory
Nature, 257(5524), 325-327 (1975-09-25)
S Nakagawa et al.
Differentiation; research in biological diversity, 29(3), 284-288 (1985-01-01)
The effects were examined of biosynthetic human epidermal growth factor (Bh-EGF) produced from cloned E. coli on DNA synthesis and all divisions of 13 different kinds of primary and established cell lines. Primary cultures of mammary epithelia, hepatocytes and stomach

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service