Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11210238001

Roche

DIG Glycan Differentiation Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100

usage

sufficient for 25 blots (10 cm x 10 cm each)

manufacturer/tradename

Roche

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

shipped in

wet ice

storage temp.

2-8°C

General description

Characterize carbohydrate moieties of glycoproteins or other, correspondingly fixed, glycoconjugates.

Application

The DIG Glycan Differentiation Kit is used to analyze carbohydrate structures by the specific binding of selected lectins. In addition to the differentiation of glycoproteins bound to nitrocellulose membranes, the lectins are also suited for the identification of carbohydrate structures on tissue sections.[1][2]

Packaging

1 kit containing 13 components.

Quality

Function tests: With the lectins GNA, SNA, MAA, and DSA 0.1 μg, and with the lectin PNA 1 μg of the control glycoproteins are detectable after a 1-min incubation in the staining solution.

Principle

Detection of digoxigenin-labeled lectins bound to specific carbohydrate structures on blots. Detection is performed using anti-digoxigenin alkaline-phosphatase conjugates. The digoxigenin-labeled lectins enable differentiation between complex and high-mannose chains, differentiation between a(2,3) and a(2,6) linkage of terminal sialic acids, and identification of the core disaccharide Galß(1,3)GalNAc of O-glycans.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • Lectin GNA, digoxigenin-labeled

  • Lectin SNA, digoxigenin-labeled

  • Lectin MAA, digoxigenin-labeled

  • Lectin PNA, digoxigenin-labeled

  • Lectin DSA, digoxigenin-labeled

  • Anti-digoxigenin-AP

  • NBT/BCIP Solution

  • Carboxypeptidase Y, Control Glycoprotein

  • Transferrin, Control Glycoprotein, HIV tested

  • Fetuin, Control Glycoprotein

  • Asialofetuin, Control Glycoprotein

  • Blocking Reagent

  • Ponceau S solution

See All (13)

signalword

Danger

Hazard Classifications

Acute Tox. 3 - Acute Tox. 4 - Acute Tox. 4 Oral - Aquatic Chronic 2 Inhalation - Resp. Sens. 1 Dermal - Skin Sens. 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lifei Yang et al.
Journal of virology, 86(14), 7662-7676 (2012-05-04)
The development of a successful vaccine against human immunodeficiency virus type 1 (HIV-1) likely requires immunogens that elicit both broadly neutralizing antibodies against envelope spikes and T cell responses that recognize multiple viral proteins. HIV-1 virus-like particles (VLP), because they
Pablo Manfredi et al.
Molecular microbiology, 81(4), 1050-1060 (2011-07-19)
Capnocytophaga canimorsus are commensal Gram-negative bacteria from dog's mouth that cause rare but dramatic septicaemia in humans. C. canimorsus have the unusual property to feed on cultured mammalian cells, including phagocytes, by harvesting the glycan moiety of cellular glycoproteins. To
Monica Lopes-Ferreira et al.
Toxins, 14(1) (2022-01-21)
TmC4-47.2 is a toxin with myotoxic activity found in the venom of Thalassophryne maculosa, a venomous fish commonly found in Latin America whose envenomation produces an injury characterized by delayed neutrophil migration, production of major pro-inflammatory cytokines, and necrosis at
Francesco Renzi et al.
mBio, 6(2), e02507-e02507 (2015-03-05)
Capnocytophaga canimorsus is known to form two kinds of cells on blood agar plates (coccoid and bacillary), evoking phase variation. When grown in coculture with animal cells these bacteria appeared only as bacilli, but in the presence of vancomycin they
Frédéric Lauber et al.
mBio, 7(5) (2016-11-01)
Bacteria of the phylum Bacteroidetes, including commensal organisms and opportunistic pathogens, harbor abundant surface-exposed multiprotein membrane complexes (Sus-like systems) involved in carbohydrate acquisition. These complexes have been mostly linked to commensalism, and in some instances, they have also been shown

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service