Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11102982001

Roche

Mro I (Acc III)

from Micrococcus roseus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105600

biological source

bacterial (Micrococcus roseus)

form

solution

packaging

pkg of 100 U (1 - 5 U/μl)

manufacturer/tradename

Roche

parameter

37 °C optimum reaction temp.

shipped in

dry ice

storage temp.

−20°C

General description

Restiction endonuclease Mro I (Acc III) recognizes the sequence TCCGGA.[1]

Specificity

Recognition sites: T↓*CCGG°A
T*CCGG°A
Restriction site: T↓*CCGG°A
T↓*CCGG°A
Heat inactivation: There is no information available whether or not Mvn I can be heat inactivated.

DNA Profile

Number of cleavage sites on different DNAs
  • λ: 24
  • φX174: 0
  • Ad2: 8
  • M13mp7: 0
  • pBR322: 1
  • pBR328: 1
  • pUC18: 0
  • SV40: 0

Unit Definition

One unit is the enzyme activity that completely cleaves 1 μg λDNA in one hour at +37 °C in a total volume of 25 μl (1x) SuRE/Cut Buffer A.

Analysis Note

SuRE/Cut Buffer System
The buffer in bold is recommended for optimal activity
  • A: 100%
  • B: 0-10%
  • H: 0-10%
  • L: 50-75%
  • M: 50-75%

Activity in PCR buffer: Not tested

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • Enzyme Solution

  • SuRE/Cut Buffer A 10x concentrated

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Inagaki et al.
Bioscience, biotechnology, and biochemistry, 57(10), 1716-1721 (1993-10-01)
A restriction endonuclease, Aor13HI, an isoschizomer of BspMII, was purified to homogeneity from cell extracts of Acidiphilium organovorum strain 13H. The enzyme has a molecular mass of 60,000 daltons and consists of two subunits identical in molecular mass of 30,000

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service