10411523001
Roche
Luciferase
from Photinus pyralis (American firefly)
About This Item
Recommended Products
form
lyophilized (recrystallized twice)
specific activity
~8 units/g protein (At 25 °C with 0.1 mM luciferin and 0.7 mM ATP as the substrates (0.1 M Tris-acetate, pH 7.75).)
packaging
pkg of 1 mg
manufacturer/tradename
Roche
optimum pH
7.5
shipped in
wet ice
General description
Application
Biochem/physiol Actions
Quality
Sequence
Preparation Note
Store the reconstituted solution in aliquots at -15 to -25 °C.
Avoid repeated freezing and thawing.
Information Note: Do not store in plastic vessels.
Reconstitution
months when stored frozen in portions of approximately 50 μl. Diluted solutions are unstable and should be prepared directly before use and should be kept in an ice-bath.
One microgram luciferase corresponds to approximately 107 arbitrary luciferase units.
Storage and Stability
Other Notes
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
does not flash
flash_point_c
does not flash
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service