10107735001
Roche
NADH
100% (dry weight), amorphous powder, pkg of 500 mg
Synonym(s):
β-Nicotinamide adenine dinucleotide, reduced disodium salt hydrate, β-DPNH, β-NADH, DPNH, Diphosphopyridine nucleotide, reduced form, NADH
About This Item
Recommended Products
description
Grade I, disodium salt
assay
100% (dry weight)
form
amorphous powder
mol wt
Mr 665.4 (NADH)
Mr 709.4 (NADH-Na2)
packaging
pkg of 500 mg
manufacturer/tradename
Roche
pH
7.5 (100 mg/mL in water, ±0.5)
7.5 ± 0.5 (100 mg/mL in water)
λmax
260 and 340 nm
shipped in
wet ice
storage temp.
2-8°C
SMILES string
O.[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]2O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)n4cnc5c(N)ncnc45)[C@@H](O)[C@H]2O
InChI
1S/C21H29N7O14P2.2Na.H2O/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33;;;/h1,3-4,7-8,10-11,13-16,20-21,29-32H,2,5-6H2,(H2,23,33)(H,34,35)(H,36,37)(H2,22,24,25);;;1H2/q;2*+1;/p-2/t10-,11-,13-,14-,15-,16-,20-,21-;;;/m1.../s1
InChI key
FWBNCIFELNNJCX-UHOVGGJYSA-L
Looking for similar products? Visit Product Comparison Guide
Application
Biochem/physiol Actions
As a reagent, NADH can be used in enzyme cycling assays to amplify detection of activity of biologically relevant enzymes or metabolites present in low concentrations.
Packaging
Quality
Organic solvents: <4% (GC)
Preparation Note
Analysis Note
Other Notes
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service