Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

10105678001

Roche

Glutathione Reductase (GR)

from yeast

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352200
Product 10105678001 is not currently sold in your country. Contact Technical Service

biological source

yeast

form

suspension

specific activity

~120 units/mg protein (At 25 °C with oxidized glutathione as the substrate; standardized with BSA.)

packaging

pkg of 1 mL (5 mg)

manufacturer/tradename

Roche

storage temp.

2-8°C

General description

Glutathione:NADP+ oxidoreductase

Application

Glutathione reductase (GR) has been used for the reduction of GSSG (glutathione disulfide) to GSH (glutathione).[1]

Biochem/physiol Actions

Glutathione Reductase (GR) is a NADPH-dependent flavoprotein that catalyzes the reduction of glutathione disulfide (GSSG) to glutathione (GSH). It is involved in the maintenance of glutathione levels, which is important for oxidation–reduction processes, detoxication and several other cellular functions.[2] In yeast, GR protects the cells from oxidative stress and is transcriptionally regulated by yAP-1 (yes-associated protein 1) protein.[3]

Quality

Contaminants: <0.01% NADPH oxidase and 6-PGDH each, <0.1% G6P-DH

Physical form

Suspension in 3.2 M ammonium sulfate solution, pH approximately 6

Preparation Note

The ammonium sulfate suspension is stable at 2 to 8 °C until the expiration date printed on the label. It may be used directly in assays for glutathione.

Activator: EDTA prevents the formation of enzyme aggregates. Phosphate stimulates the NADH-dependent reaction.

Other Notes

For life science research only. Not for use in diagnostic procedures.

related product

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C M Grant et al.
Molecular microbiology, 21(1), 171-179 (1996-07-01)
Glutathione (GSH) is an abundant cellular thiol which has been implicated in many cellular processes including protection against xenobiotics, carcinogens and free radicals. Utilization of GSH in both enzymic and non-enzymic defence mechanisms results in its conversion to the oxidized
Glutathione reductase.
I Carlberg et al.
Methods in enzymology, 113, 484-490 (1985-01-01)
Trent E Tipple et al.
Methods in molecular biology (Clifton, N.J.), 889, 315-324 (2012-06-07)
We present two different methods for determining levels of glutathione in complex biological samples and plasma. The DTNB/GR enzyme recycling method is sensitive and requires no specialized equipment. The HPLC method is particularly useful for situations in which sample amounts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service