Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z742600

Whatman® anion exchange cellulose

DE52

Sign Into View Organizational & Contract Pricing


About This Item

packaging

pkg of 500 g

manufacturer/tradename

Whatman 4057-050
Whatman Article No. 28419498 (US reference)

Looking for similar products? Visit Product Comparison Guide

General description

The DE weak (charge dependent on pH) anion exchangers are based on the diethylaminoethyl (DEAE) tertiary amine functional group. QA52 is a strong (charge independent of pH) anion exchange medium, containing quaternary amine groups.

DE23 dry fibrous DEAE cellulose - Allows fast flow rates especially after fines removal; suitable for negatively charged biopolymers.

DE32 dry microgranular DEAE cellulose - Similar performance characteristics after precycling as DE52.

DE52 preswollen microgranular DEAE cellulose - Probably the most widely used DEAE cellulose in the world; used for biopolymers with low to high negative charges; exhibits excellent resolution with good flow rates.

DE53 preswollen microgranular DEAE cellulose - Partially quaternized DEAE anion exchanger, highly substituted and with higher capacity than DE52; can be used in series with DE52 media.

QA52 preswollen microgranular QA cellulose - A strongly basic, quaternary amine-bearing anion exchange medium, moderately substituted, with high protein capacity. Fully ionized, bears constant charge under all pH conditions; excellent for high pH applications.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service