Z276529
FT-IR Spectral Libraries
Nicolet®/Aldrich Raman Condensed Phase Library
About This Item
General description
• Contain spectral data from 4,000 to 450 cm-1
• Best used when searching compounds with significant spectral differences
• Generally not suitable when using spectral subtraction or other precision spectral identification techniques
Other Notes
• Alcohols and phenols
• Aldehydes and ketones
• Carboxylic acids, esters, lactones, and anhydrides
• Polymers
• Solvents
• More
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)