About This Item
Recommended Products
grade
Kosher
natural
reg. compliance
FCC
assay
≥98%
impurities
15-25% 2-methylbutyl salicylate
refractive index
n20/D 1.507 (lit.)
bp
277-278 °C (lit.)
density
1.05 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
SMILES string
CC(C)CCOC(=O)c1ccccc1O
InChI
1S/C12H16O3/c1-9(2)7-8-15-12(14)10-5-3-4-6-11(10)13/h3-6,9,13H,7-8H2,1-2H3
InChI key
PMGCQNGBLMMXEW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Biochem/physiol Actions
Other Notes
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
flash_point_f
230.0 °F - closed cup
flash_point_c
110 °C - closed cup
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service