Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W521507

Sigma-Aldrich

Cedryl acetate

Synonym(s):

(+)-Cedryl acetate, (3R,3aS,6R,7R,8aS)-Octahydro-3,6,8,8-tetramethyl-1H-3a,7-methanoazulen-6-ol-6-acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H28O2
CAS Number:
Molecular Weight:
264.40
Beilstein/REAXYS Number:
2052432
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.171

biological source

synthetic

refractive index

n20/D 1.499 (lit.)

bp

>200 °C (lit.)

density

0.999 g/mL at 25 °C (lit.)

organoleptic

woody

SMILES string

C[C@@H]1CC[C@H]2C(C)(C)[C@H]3C[C@@]12CC[C@@]3(C)OC(C)=O

InChI

1S/C17H28O2/c1-11-6-7-13-15(3,4)14-10-17(11,13)9-8-16(14,5)19-12(2)18/h11,13-14H,6-10H2,1-5H3/t11-,13+,14-,16-,17+/m1/s1

InChI key

HQKQRXZEXPXXIG-DTWJZALFSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service