Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W517909

Sigma-Aldrich

Diethyl phthalate

≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(CO2C2H5)2
CAS Number:
Molecular Weight:
222.24
Beilstein/REAXYS Number:
1912500
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Pricing and availability is not currently available.

biological source

synthetic

vapor density

7.66 (vs air)

vapor pressure

1 mmHg ( 100 °C)

assay

≥99.5%

autoignition temp.

854 °F

expl. lim.

0.75 %, 187 °F

refractive index

n20/D 1.502 (lit.)

bp

298-299 °C (lit.)

mp

−3 °C (lit.)

density

1.12 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

organoleptic

faint

SMILES string

CCOC(=O)c1ccccc1C(=O)OCC

InChI

1S/C12H14O4/c1-3-15-11(13)9-7-5-6-8-10(9)12(14)16-4-2/h5-8H,3-4H2,1-2H3

InChI key

FLKPEMZONWLCSK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

338.0 °F - closed cup

flash_point_c

170 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raphael J Witorsch et al.
Critical reviews in toxicology, 40 Suppl 3, 1-30 (2010-10-12)
This article reviews laboratory and epidemiological research into the endocrine disruptive effects of components of personal care products, namely, phthalate esters, parabens, ultraviolet (UV) filters, polycyclic musks, and antimicrobials. High doses of phthalates in utero can produce “phthalate syndrome,” demasculinizing
Kashappa-Goud H Desai et al.
Journal of controlled release : official journal of the Controlled Release Society, 165(1), 62-74 (2012-10-30)
Herein, we describe the detailed development of a simple and effective method to microencapsulate vaccine antigens in poly(lactic-co-glycolic acid) (PLGA) by simple mixing of preformed active self-microencapsulating (SM) PLGA microspheres in a low concentration aqueous antigen solution at modest temperature
A M Api
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 39(2), 97-108 (2001-03-27)
Diethyl phthalate (DEP; CAS No. 84-66-2) has many industrial uses, as a solvent and vehicle for fragrance and cosmetic ingredients and subsequent skin contact. This review focuses on its safety in use as a solvent and vehicle for fragrance and
Lee-Ju Cheng et al.
Aquatic toxicology (Amsterdam, Netherlands), 109, 166-175 (2011-10-28)
The toxicity and effects of diethyl phthalate (DEP), a potent allelochemical, on the growth of greater duckweed were studied. Biochemical analyses and physiological methods were combined to investigate oxidative stress, adverse effects and their mechanisms in greater duckweeds grown in
Maik Hadorn et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(50), 20320-20325 (2012-11-24)
Higher-order structures that originate from the specific and reversible DNA-directed self-assembly of microscopic building blocks hold great promise for future technologies. Here, we functionalized biotinylated soft colloid oil-in-water emulsion droplets with biotinylated single-stranded DNA oligonucleotides using streptavidin as an intermediary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service