Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W517003

Sigma-Aldrich

1-Hepten-3-ol

≥97%

Synonym(s):

Butyl vinyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(OH)CH=CH2
CAS Number:
Molecular Weight:
114.19
Beilstein/REAXYS Number:
1720340
EC Number:
MDL number:
UNSPSC Code:
12164502
Flavis number:
2.155

assay

≥97%

refractive index

n20/D 1.433 (lit.)

bp

153-156 °C (lit.)

density

0.836 g/mL at 20 °C
0.836 g/mL at 25 °C (lit.)

SMILES string

CCCCC(O)C=C

InChI

1S/C7H14O/c1-3-5-6-7(8)4-2/h4,7-8H,2-3,5-6H2,1H3

InChI key

PZKFYTOLVRCMOA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

128.8 °F - closed cup

flash_point_c

53.8 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service