Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W515000

Sigma-Aldrich

Butyl benzoate

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOCH2CH2CH2CH3
CAS Number:
Molecular Weight:
178.23
Beilstein/REAXYS Number:
1867073
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.779

biological source

synthetic

grade

Halal
Kosher

assay

≥98%

refractive index

n20/D 1.498 (lit.)

bp

249 °C (lit.)

density

1.01 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

fruity

SMILES string

CCCCOC(=O)c1ccccc1

InChI

1S/C11H14O2/c1-2-3-9-13-11(12)10-7-5-4-6-8-10/h4-8H,2-3,9H2,1H3

InChI key

XSIFPSYPOVKYCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged in glass bottles

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

222.8 °F - closed cup

flash_point_c

106 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suvi-Katriina Ruokonen et al.
Journal of chromatography. A, 1405, 178-187 (2015-06-15)
The effect of three phosphonium and imidazolium ionic liquids (ILs) on the interaction between liposomes and common pharmaceuticals found in wastewaters was studied. The liposomes comprised zwitterionic phosphatidyl choline and negatively charged phosphatidyl glycerol. A set of common cationic, anionic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service