W511501
2-Methoxythiophenol
≥97%, FG
Synonym(s):
2-Mercaptoanisole, 2-Methoxybenzenethiol, Thioguaiacol
About This Item
grade
FG
reg. compliance
EU Regulation 1334/2008 & 178/2002
assay
≥97%
refractive index
n20/D 1.591 (lit.)
bp
99 °C/8 mmHg (lit.)
density
1.152 g/mL at 25 °C (lit.)
documentation
see Safety & Documentation for available documents
food allergen
no known allergens
SMILES string
COc1ccccc1S
InChI
1S/C7H8OS/c1-8-6-4-2-3-5-7(6)9/h2-5,9H,1H3
InChI key
DSCJETUEDFKYGN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.
replaced by
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
204.8 °F - closed cup
flash_point_c
96 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service