Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W508330

Sigma-Aldrich

cis-2-Hexen-1-ol

≥90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH=CHCH2OH
CAS Number:
Molecular Weight:
100.16
Beilstein/REAXYS Number:
1719708
EC Number:
MDL number:
Flavis number:
2.156
Pricing and availability is not currently available.

vapor density

>1 (vs air)

assay

≥90%

impurities

<10% trans isomer

refractive index

n20/D 1.441 (lit.)

bp

166 °C (lit.)

density

0.847 g/mL at 25 °C (lit.)

SMILES string

[H]\C(CO)=C(/[H])CCC

InChI

1S/C6H12O/c1-2-3-4-5-6-7/h4-5,7H,2-3,6H2,1H3/b5-4-

InChI key

ZCHHRLHTBGRGOT-PLNGDYQASA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

138.9 °F - closed cup

flash_point_c

59.4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service