Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W507970

Sigma-Aldrich

1,5-Pentanedithiol

Synonym(s):

Pentamethylene dimercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HS(CH2)5SH
CAS Number:
Molecular Weight:
136.28
Beilstein/REAXYS Number:
1732335
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Pricing and availability is not currently available.

refractive index

n20/D 1.519 (lit.)

bp

107-108 °C/15 mmHg (lit.)

mp

−72 °C (lit.)

density

1.016 g/mL at 25 °C (lit.)

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

SMILES string

SCCCCCS

InChI

1S/C5H12S2/c6-4-2-1-3-5-7/h6-7H,1-5H2

Inchi Key

KMTUBAIXCBHPIZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

203.0 °F - closed cup

flash_point_c

95 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service