Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W507210

Sigma-Aldrich

Phenyl salicylate

≥99%

Synonym(s):

Salol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CO2C6H5
CAS Number:
Molecular Weight:
214.22
Beilstein/REAXYS Number:
393969
EC Number:
MDL number:
UNSPSC Code:
12164502
Flavis number:
9.689

assay

≥99%

bp

172-173 °C/12 mmHg (lit.)

mp

41-43 °C (lit.)

SMILES string

Oc1ccccc1C(=O)Oc2ccccc2

InChI

1S/C13H10O3/c14-12-9-5-4-8-11(12)13(15)16-10-6-2-1-3-7-10/h1-9,14H

InChI key

ZQBAKBUEJOMQEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Mild and sweet, fruity balsamic odor

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service