W503800
3-Decanone
≥97%
Synonym(s):
Ethyl heptyl ketone
About This Item
biological source
synthetic
assay
≥97%
refractive index
n20/D 1.4240 (lit.)
bp
204-205 °C (lit.)
mp
−4-−3 °C (lit.)
density
0.825 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
organoleptic
fatty; green; orange; floral
SMILES string
CCCCCCCC(=O)CC
InChI
1S/C10H20O/c1-3-5-6-7-8-9-10(11)4-2/h3-9H2,1-2H3
InChI key
XJLDYKIEURAVBW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
143.6 °F - closed cup
flash_point_c
62 °C - closed cup
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service