Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W501204

Sigma-Aldrich

Propyl tiglate

≥98%

Synonym(s):

Propyl trans-2-methyl-2-butenoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=C(CH3)CO2CH2CH2CH3
CAS Number:
Molecular Weight:
142.20
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:

biological source

synthetic

assay

≥98%

refractive index

n20/D 1.4371 (lit.)

bp

176-180 °C (lit.)

density

0.904 g/mL at 25 °C (lit.)

SMILES string

CCCOC(=O)\C(C)=C\C

InChI

1S/C8H14O2/c1-4-6-10-8(9)7(3)5-2/h5H,4,6H2,1-3H3/b7-5+

InChI key

RZWMDOQSXWAAMC-FNORWQNLSA-N

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

136.4 °F - closed cup

flash_point_c

58 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service