Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W430400

Sigma-Aldrich

trans-2-Decen-1-ol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6CH=CHCH2OH
CAS Number:
Molecular Weight:
156.27
FEMA Number:
4304
EC Number:
MDL number:
UNSPSC Code:
12164502

biological source

synthetic

assay

97%

refractive index

n20/D 1.451 (lit.)

bp

231-232 °C (lit.)

density

0.844 g/mL at 25 °C (lit.)

SMILES string

CCCCCCC\C=C\CO

InChI

1S/C10H20O/c1-2-3-4-5-6-7-8-9-10-11/h8-9,11H,2-7,10H2,1H3/b9-8+

InChI key

QOPYYRPCXHTOQZ-CMDGGOBGSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service