Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W423901

Sigma-Aldrich

Isobutylamine

99%

Synonym(s):

1-Amino-2-methylpropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2NH2
CAS Number:
Molecular Weight:
73.14
FEMA Number:
4239
Beilstein/REAXYS Number:
385626
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
11.002

biological source

synthetic

assay

99%

refractive index

n20/D 1.397 (lit.)

bp

64-71 °C (lit.)

mp

−85 °C (lit.)

density

0.736 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

organoleptic

fishy

SMILES string

CC(C)CN

InChI

1S/C4H11N/c1-4(2)3-5/h4H,3,5H2,1-2H3

Inchi Key

KDSNLYIMUZNERS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Flam. Liq. 2 - Skin Corr. 1A

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

15.8 °F - closed cup

flash_point_c

-9 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chun-Zhao Liu et al.
Journal of agricultural and food chemistry, 54(22), 8456-8460 (2006-10-26)
Inoculation of leaf explants of Echinacea purpurea (Moench) with Agrobacterium rhizogenes induced hairy roots with the capacity to produce biologically active caffeic acid derivatives (CADs), especially cichoric acid. The kinetics of growth, the uptake of macronutrients, and the accumulation of
L Wang et al.
Journal of immunology (Baltimore, Md. : 1950), 167(11), 6195-6201 (2001-11-21)
Whereas cytokine production in alphabeta T cells is rapidly regulated by exposure to peptide Ag, the mechanisms regulating cytokine production by gammadelta T cells are unknown. In this study, we demonstrate that human Vgamma2Vdelta2 T cells produce IFN-gamma and TNF-alpha
R J Parry et al.
Journal of bacteriology, 179(2), 409-416 (1997-01-01)
The flavoprotein isobutylamine N-hydroxylase (IBAH) catalyzes the oxidation of isobutylamine to isobutylhydroxylamine, a key step in the biosynthesis of the azoxy antibiotic valanimycin. By using oligonucleotide primers designed from peptide sequence information derived from native IBAH, a fragment of the
[Biosynthesis of nitrosopiperidine from isobutylamine and NaNo2 by some fungi].
C A Ji et al.
Wei sheng wu xue bao = Acta microbiologica Sinica, 27(1), 78-82 (1987-03-01)
Kimiyasu Isobe et al.
Journal of chromatography. A, 1144(1), 85-89 (2006-11-23)
A convection interaction media (trade name CIM, BIA Separation, Ljubljana, Slovenia) isobutyl monolithic disc was prepared by incubating a CIM epoxy monolithic disc with isobutylamine, and it was then applied to the purification of secondary alcohol dehydrogenase (S-ADH) and primary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service