Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W410601

Sigma-Aldrich

2,5-Dimethylfuran

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O
CAS Number:
Molecular Weight:
96.13
FEMA Number:
4106
Beilstein/REAXYS Number:
106449
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.029

vapor density

3.31 (vs air)

assay

≥99%

refractive index

n20/D 1.441 (lit.)

bp

92-94 °C (lit.)

mp

−62 °C (lit.)

density

0.903 g/mL at 25 °C (lit.)

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

SMILES string

Cc1ccc(C)o1

InChI

1S/C6H8O/c1-5-3-4-6(2)7-5/h3-4H,1-2H3

InChI key

GSNUFIFRDBKVIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

33.8 °F

flash_point_c

1 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service