Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W404901

Sigma-Aldrich

Syringaldazine

Synonym(s):

4-Hydroxy-3,5-dimethoxybenzaldehyde azine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[HOC6H2(OCH3)2CH=N-]2
CAS Number:
Molecular Weight:
360.36
FEMA Number:
4049
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Pricing and availability is not currently available.

biological source

synthetic

mp

209-210 °C (lit.)

application(s)

flavors and fragrances

SMILES string

COc1cc(\C=N\N=C\c2cc(OC)c(O)c(OC)c2)cc(OC)c1O

InChI

1S/C18H20N2O6/c1-23-13-5-11(6-14(24-2)17(13)21)9-19-20-10-12-7-15(25-3)18(22)16(8-12)26-4/h5-10,21-22H,1-4H3/b19-9+,20-10+

Inchi Key

YARKTHNUMGKMGS-LQGKIZFRSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Flame

signalword

Warning

hcodes

pcodes

Hazard Classifications

Flam. Sol. 2

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Rajendiran et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 68(3), 894-904 (2007-02-24)
The absorption and fluorescence spectra of syringaldazine (SYAZ) has been recorded in solvents of different polarity, pH and beta-cyclodextrin (beta-CD) and compared with syringaldehyde (SYAL). The inclusion complex of SYAZ with beta-CD is investigated by UV-vis, fluorimetry, AM 1, FT-IR
Qiu Weihua et al.
Bioresource technology, 99(13), 5480-5484 (2007-12-22)
Mycelia Sterilia YY-5, an entophytic fungus, was isolated from Rhus chinensis Mill and its extracellular enzyme had a higher laccase activity (MS-Lac). After been purified by anion exchange and gel filtration chromatography, MS-Lac, which had a molecular mass of 45
M Lorenzo et al.
Chemosphere, 60(8), 1124-1128 (2005-07-05)
Due to the numerous biotechnological applications of laccase enzyme, it is essential to know the influence of different agents usually present in the natural environment on its enzymatic action, especially for in situ treatment technologies. In the present work, a
Makoto Hattori et al.
Journal of insect physiology, 51(12), 1359-1365 (2005-10-12)
The activity and composition of leafhopper saliva are important in interactions with the host rice plant, and it may play a physiological role in detoxifying toxic plant substances or ingesting sap. We have characterized diphenoloxidase in the salivary glands of
Wojciech Nogala et al.
Bioelectrochemistry (Amsterdam, Netherlands), 79(1), 101-107 (2010-01-26)
Electrodes modified with sol-gel encapsulated laccase (isolated from Cerrenaunicolor) exhibiting mediated or mediatorless bioelectrocatalytic dioxygen reduction activity were inspected using confocal laser scanning microscopy, atomic force microscopy and scanning electrochemical microscopy. Potential-driven leaching of the redox mediator 2,2'-azino-bis(3-ethylbenzothiazoline-6-sulfonate) from carbon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service